Learn More
enQuireBio™ TranslationBlocker Human Apoptosis regulator BAX siRNA
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
Specifications
Specifications
| Accession Number | NP_004315 |
| Cell Lines Tested | KAT-4 Human Thyroid Carcinoma Cells and Human T Cells. |
| Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
| Gene Symbol | BAX |
| Gene ID (Entrez) | 581 |
| Product Type | siRNA |
| Quantity | 10 nmol |
| Synonym | BCL2L4 |
| Target | Human |
| Form | Lyophilized |