Learn More
enQuireBio™ TranslationBlocker Human ApoE siRNA
Specifications
Specifications
| Accession Number | NP_000032 |
| Cell Lines Tested | Ovcar3 Human Ovarian Cancer Cells. |
| Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
| Gene Symbol | APOE |
| Gene ID (Entrez) | 348 |
| Product Type | siRNA |
| Quantity | 10 nmol |
| Synonym | AD2, APO-E, LPG, LDLCQ5, ApoE4, ApoE, Apolipoprotein E |
| Target | Human |
| Form | Lyophilized |