Learn More
enQuireBio™ TranslationBlocker Human Androgen receptor siRNA
Product Code. 16099221
Change view
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
Product Code. Quantity unitSize
16099221 2 nmol 2nmol
16089221 10 nmol 10nmol
2 options
Product Code. 16099221
Brand: enQuireBio™ QX42nmol
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Blocks transcription and translation of Androgen receptor from Human.
Specifications
Accession Number NP_000035
Cell Lines Tested LNCaP and LNCaP-Rf Androgen Dependent and Independent Metastatic Cells..
Content And Storage Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
Gene Symbol AR
Gene ID (Entrez) 367
Product Type siRNA
Quantity 2 nmol
Synonym SMAX1, AR8, KD, SBMA, HUMARA, DHTR, HYSP1, TFM, AIS, NR3C4
Target Human
Form Lyophilized