missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human Androgen receptor siRNA
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
Specifications
Specifications
| Accession Number | NP_000035 |
| Cell Lines Tested | LNCaP and LNCaP-Rf Androgen Dependent and Independent Metastatic Cells.. |
| Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
| Gene Symbol | AR |
| Gene ID (Entrez) | 367 |
| Product Type | siRNA |
| Quantity | 10 nmol |
| Synonym | SMAX1, AR8, KD, SBMA, HUMARA, DHTR, HYSP1, TFM, AIS, NR3C4 |
| Target | Human |
| Form | Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction