Learn More
enQuireBio™ TranslationBlocker Human Amyloid beta A4 protein siRNA
Specifications
Specifications
| Accession Number | NP_000475 |
| Cell Lines Tested | HEK 293. |
| Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
| Gene Symbol | APP |
| Gene ID (Entrez) | 351 |
| Product Type | siRNA |
| Quantity | 2 nmol |
| Synonym | CTFgamma, ABETA, AD1, PN2, ABPP, CVAP, AAA, APPI, PN-II, ab |
| Target | Human |
| Form | Lyophilized |