Learn More
enQuireBio™ TranslationBlocker Human Alpha-synuclein siRNA
Product Code. 16059181
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
Product Code. 16059181
Brand: enQuireBio™ QX22nmol
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Blocks transcription and translation of Alpha-synuclein from Human.
Specifications
Accession Number NP_000336
Cell Lines Tested SH-SY5Y.
Content And Storage Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
Gene Symbol SNCA
Gene ID (Entrez) 6622
Product Type siRNA
Quantity 2 nmol
Synonym PD1, NACP, PARK1, PARK4, SNCA
Target Human
Form Lyophilized