Learn More
enQuireBio™ TranslationBlocker Human ALK tyrosine kinase receptor / CD246 siRNA
Product Code. 16049161
Change view
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
Product Code. Quantity unitSize
16049161 10 nmol 10nmol
16059161 2 nmol 2nmol
2 options
Product Code. 16049161
Brand: enQuireBio™ QX110nmol
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Blocks transcription and translation of ALK tyrosine kinase receptor / CD246 from Human.
Spezifikation
Accession Number NP_004295
Cell Lines Tested SK-N-MC and NB-39-nu Neuroblastoma Cell Lines.
Content And Storage Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
Gene Symbol ALK
Gene ID (Entrez) 238
Product Type siRNA
Quantity 10 nmol
Synonym CD246, NBLST3, Anaplastic lymphoma kinase
Target Human
Form Lyophilized