Learn More
BRAND™ Transferpettor™ Pipetter Station
Product Code. p-8555018 Shop All Brand Products
Click to view available options
Positions:
2
4
Unit Size:
Each
Product Code. 10463911
Brand: BRAND™ 702890
Only null left
Add to basket
Add to basket
Provides handy and easily accessible storage for Transferpettor™ instruments and accessories.
  • Choose to accommodate 2 or 4 instruments up to 200 μL
  • Holds pipettes upright
  • Transferpettors sold separately.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Pipette Holder
Autoclavable Not Autoclavable
Color White
Holds Manual Pipettes
Material Plastic
Positions 2
Shape Rectangular
Shelves None
Stackable No
Sterility Sterile
For Use With (Equipment) Transferpettor pipetting devices
Width (Metric) 19.05 cm
Width (English) 7.5 in.
Height (Metric) 12.7 cm
Height (English) 5 in.
Depth (Metric) 22.86 cm