Learn More
BRAND™ Transferpette™ S Pipette Bench Rack
Product Code. 16415999 Shop All Brand Products
Change view
Click to view available options
Holds:
Pipettes (6)
Unit Size:
Each
Product Code. Holds unitSize
16415999 Pipettes (6) Each
1 options
Product Code. 16415999
Brand: BRAND™ 704807
Only null left
Add to basket
Add to basket
Accessory for use with Transferpette™ S single and multichannel pipettes.
  • Holds six instruments
  • Rotates for easy pipette access
  • Pipettes sold separately
TRUSTED_SUSTAINABILITY
Specifications
Description Rotates
Autoclavable Not Autoclavable
Color White
Holds Pipettes (6)
Material Plastic
Positions 6
Shape Circular
Stackable No
Sterility Non-sterile
For Use With (Equipment) Transferpette S Pipettes
Product Type Benchtop Rack
Shelves None