Learn More
Thermo Scientific™ Transfer Vessels Withdrawal Device
Product Code. 10025753 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
LN2 Transfer Vessels, For Thermo 10
LN2 Transfer Vessels, For Thermo 20
LN2 Transfer Vessels, For Thermo 30
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
10025753 LN2 Transfer Vessels, For Thermo 10 Each
10723514 LN2 Transfer Vessels, For Thermo 30 Each
12794716 LN2 Transfer Vessels, For Thermo 20 Each
3 options
Product Code. 10025753
Brand: Thermo Scientific™ AY509X5
Only null left
Add to basket
Add to basket
Easily dispense up to 8L/min. (2.12 gal./min.) without pouring using the Thermo Scientific™ Transfer Vessels Withdrawal Device.
  • Available for 10, 20 and 30L models
  • TRUSTED_SUSTAINABILITY
    Specifications
    Type LN2 Withdrawal Device, 8L/min.
    Capacity (English) 1.05 cu. ft.
    Capacity (Metric) 30 L
    For Use With (Equipment) LN2 Transfer Vessels, For Thermo 10