Learn More
Invitrogen™ Transfer Tube, Female Luer Lock, 36 inch
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
With female luer lock.
Radiation sterilized.
Radiation sterilized.
Specifications
Specifications
| Quantity | 1 Ea. |
| Sterility | Sterile |
| Content And Storage | Store at room temperature. |
| Shipping Condition | Room Temperature |
| Length (Metric) | 914.4 mm |
| Product Type | Transfer Tube |