Learn More
(+)-trans Taxifolin, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | (+)-Taxifolin |
| CAS | 480-18-2 |
| Molecular Formula | C15 H12 O7 |
| Quantity | 500 mg |
| Synonym | 4H-1-Benzopyran-4-one','2-(3,4-dihydroxyphenyl)-2,3-dihydro-3,5,7-trihydroxy-','(2R,3R)-, 4H-1-Benzopyran-4-one','2-(3,4-dihydroxyphenyl)-2,3-dihydro-3,5,7-trihydroxy-','(2R-trans)-, Flavanone','3,3',4',5,7-pentahydroxy- (8CI), (2R,3R)-2-(3,4-Dihydroxyphenyl)-2,3-dihydro-3,5,7-trihydroxy-4H-1-benzopyran-4-one, (+)-(2R,3R)-Dihydroquercetin, (+)-Dihydroquercetin, (+)-Taxifolin, (+)-trans-Taxifolin, (2R,3R)-(+)-Taxifolin, (2R,3R)-3,3',4',5,7-Pentahydroxyflavanone, (2R,3R)-Dihydroquercetin, 2,3-Dihydroquercetin, 3,5,7,3',4'-Pentahydroxyflavanone, Dihydroquercetin, Diquertin, Distylin, Flamena, Flamena D, Jikuberuchin, Lariksin, Lavitol, Taxifolin, Taxifoliol |
| InChI Formula | InChI=1S/C15H12O7/c16-7-4-10(19)12-11(5-7)22-15(14(21)13(12)20)6-1-2-8(17)9(18)3-6/h1-5,14-19,21H/t14-,15+/m0/s1 |
| SMILES | O[C@@H]1[C@H](Oc2cc(O)cc(O)c2C1=O)c3ccc(O)c(O)c3 |
| IUPAC Name | (2R,3R)-2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxy-2,3-dihydrochromen-4-one |
| Molecular Weight (g/mol) | 304.25 |
| Formula Weight | 304.0583 |