Learn More
trans-Chalcone, 97%
Description
Trans chalcone is used to prevent lung and forestomach cancer. It induces programmed cell death due to reduced mitochondrial transmembrane potential in Arabidopsis thaliana roots.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Trans chalcone is used to prevent lung and forestomach cancer. It induces programmed cell death due to reduced mitochondrial transmembrane potential in Arabidopsis thaliana roots.
Solubility
Soluble in chloroform, ether, benzene, and ethanol (slightly). Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 55°C to 57°C |
| Boiling Point | 208°C (25 mmHg) |
| Quantity | 100 g |
| Beilstein | 509985 |
| Merck Index | 14,2037 |
| Solubility Information | Soluble in chloroform,ether,benzene,and ethanol (slightly). Insoluble in water. |
| Formula Weight | 208.26 |
| Percent Purity | 97% |
| Chemical Name or Material | trans-Chalcone |
Safety and Handling
- trans-Chalcone
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contain: (E)-Chalcone
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.