Learn More
trans-4-Hydroxy-3-methoxycinnamic acid, 99%
Description
A chemopreventive analogue of caffeic acid.trans-4-Hydroxy-3-methoxycinnamic acid is used as an important raw material for the preparation of its corresponding esters by reacting with alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
A chemopreventive analogue of caffeic acid.trans-4-Hydroxy-3-methoxycinnamic acid is used as an important raw material for the preparation of its corresponding esters by reacting with alcohols.
Solubility
Soluble in alcohol and hot water.
Notes
Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 170°C to 175°C |
| Quantity | 500 g |
| Beilstein | 1371483 |
| Merck Index | 14,4062 |
| Solubility Information | Soluble in alcohol and hot water. |
| Formula Weight | 194.19 |
| Percent Purity | 99% |
| Chemical Name or Material | trans-4-Hydroxy-3-methoxycinnamic acid |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.