missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Ethylcyclohexanecarboxylic acid, 98%, Thermo Scientific™

Product Code. 11464967
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464967 1 g 1g
11474967 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11464967 Supplier Thermo Scientific Alfa Aesar Supplier No. B21897.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6833-47-2
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.23
MDL Number MFCD01076323,MFCD01568857
InChI Key UNROFSAOTBVBBT-UHFFFAOYSA-N
Synonym trans-4-ethylcyclohexanecarboxylic acid, 4-ethylcyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-ethyl, 4-trans-ethyl cyclohexane carboxylic acid, cyclohexanecarboxylic acid, 4-ethyl-, trans, 1s,4r-4-ethylcyclohexane-1-carboxylic acid, pubchem2491, acmc-209o2z, ksc352s1t, ksc352s2b
PubChem CID 1490288
IUPAC Name 4-ethylcyclohexane-1-carboxylic acid
SMILES CCC1CCC(CC1)C(O)=O

Specifications

Melting Point 48°C to 51°C
Quantity 1 g
Formula Weight 156.23
Percent Purity 98%
Chemical Name or Material trans-4-Ethylcyclohexanecarboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.