Learn More
trans-4-[[(2-Amino-5-bromophenyl)methyl]amino]-cyclohexanol, TRC
Product Code. 30010781
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010781 50 mg 50mg
30005849 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30010781 Supplier Toronto Research Chemicals Supplier No. TRCA60198550MG
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 101900-43-0
Molecular Formula C13 H19 Br N2 O
Molecular Weight (g/mol) 299.21
Synonym trans-4-[[(2-amino-5-bromophenyl)methyl]amino]cyclohexanol, Ambroxol Monobromine
IUPAC Name 4-[(2-amino-5-bromophenyl)methylamino]cyclohexan-1-ol
SMILES Nc1ccc(Br)cc1CN[C@@H]2CC[C@@H](O)CC2
Specifications
Analyte or Component Names trans-4-[((2-Amino-5-bromophenyl)methyl)amino]-cyclohexanol
Shipping Condition Room Temperature
InChI Formula InChI=1S/C13H19BrN2O/c14-10-1-6-13(15)9(7-10)8-16-11-2-4-12(17)5-3-11/h1,6-7,11-12,16-17H,2-5,8,15H2/t11-,12-
Solution Type Neat
Industry Type Forensics
Quantity 50 mg
Formula Weight 298.0681
Chemical Name or Material trans-4-[((2-Amino-5-bromophenyl)methyl)amino]-cyclohexanol