Learn More
trans-3-Hexen-1-ol, 97%
Product Code. 11358495
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11358495
Brand: Thermo Scientific Alfa Aesar L10541.14
Request Bulk or Custom Format
CAS: 928-97-2 | C6H12O | 100.161 g/mol
trans-3-Hexen-1-ol stimulates the antennae of male H. cunea moths. It is also used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
trans-3-Hexen-1-ol stimulates the antennae of male H. cunea moths. It is also used as an organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 928-97-2
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00002960
InChI Key UFLHIIWVXFIJGU-ONEGZZNKSA-N
Synonym trans-3-hexen-1-ol, e-hex-3-en-1-ol, e-3-hexen-1-ol, trans-3-hexenol, 3-hexen-1-ol, 3e-hex-3-en-1-ol, 3-hexen-1-ol, e, trans-hex-3-en-1-ol, unii-4e0nfr5b3u, 3e-hexen-1-ol
PubChem CID 5284503
IUPAC Name (E)-hex-3-en-1-ol
SMILES CCC=CCCO
Specifications
Density 0.845
Boiling Point 60°C to 61°C (12 mmHg)
Flash Point 58°C (136°F)
Odor Pleasant
Refractive Index 1.439
Quantity 25 g
UN Number UN1987
Beilstein 1719713
Merck Index 14,4700
Solubility Information Slightly soluble in water.
Formula Weight 100.16
Percent Purity 97%
Chemical Name or Material trans-3-Hexen-1-ol