missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-2-Undecenoic acid, tech. 80%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Boiling Point | 138°C to 140°C (2 mmHg) |
| Refractive Index | 1.4625 |
| Quantity | 5 g |
| Beilstein | 1722519 |
| Formula Weight | 184.28 |
| Percent Purity | 80% |
| Grade | Technical |
| Chemical Name or Material | trans-2-Undecenoic acid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur