Learn More
trans-2-Pentene, 99%
Product Code. 11377416
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11377416 5 g 5g
11367416 1 g 1g
2 options
Product Code. 11377416
Brand: Thermo Scientific Alfa Aesar L09421.06
Request Bulk or Custom Format
CAS: 646-04-8 | C5H10 | 70.135 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 646-04-8
Molecular Formula C5H10
Molecular Weight (g/mol) 70.135
MDL Number MFCD00009384
InChI Key QMMOXUPEWRXHJS-HWKANZROSA-N
Synonym trans-2-pentene, e-2-pentene, 2-pentene, 3-pentene, 2-pentene, e, sym-methylethylethylene, e-pent-2-ene, beta-n-amylene, trans-beta-amylene, 2-pentene, 2e
PubChem CID 5326161
IUPAC Name (E)-pent-2-ene
SMILES CCC=CC
Specifications
Melting Point -136°C
Density 0.649
Boiling Point 36°C to 37°C
Flash Point −45°C (−49°F)
Odor Petroleum-like
Refractive Index 1.379
Quantity 5 g
UN Number UN3295
Beilstein 1718795
Merck Index 14,7123
Formula Weight 70.14
Percent Purity 99%
Chemical Name or Material trans-2-Pentene