Learn More
trans-2-Hexenyl acetate, 98%
Product Code. 11443068
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11443068 10 g 10g
11453068 50 g 50g
11463068 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11443068 Supplier Thermo Scientific Alfa Aesar Supplier No. A18786.09
 Restricted Item
In Stock
Add to basket
CAS: 2497-18-9 | C8H14O2 | 142.198 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2497-18-9
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00009474
InChI Key HRHOWZHRCRZVCU-AATRIKPKSA-N
Synonym z-hex-2-enyl acetate, unii-pt792bq57d, 2-hexen-1-ol, acetate, z, z-2-hexenyl acetate, fema no. 2564, 2-hexen-1-yl acetate, trans, 2-hexen-1-ol, acetate, 2e, 2-hexenyl acetate, trans, hexenyl acetate, 2z
PubChem CID 2733294
IUPAC Name [(E)-hex-2-enyl] acetate
SMILES CCCC=CCOC(=O)C
Specifications
Boiling Point 165°C to 166°C
Flash Point 58°C (136°F)
Quantity 10 g
UN Number UN3272
Beilstein 1721851
Formula Weight 142.2
Percent Purity 98%
Chemical Name or Material trans-2-Hexenyl acetate
compliance-icons compliance-icons
Product Identifier
  • trans-2-Hexenyl acetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: trans-2-Hexenyl acetate