Learn More
trans-1-Fluoro-4-(4-n-pentylcyclohexyl)benzene, 97%, Thermo Scientific™
Product Code. 15471996
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15471996 5 g 5g
15461996 1 g 1g
2 options
Product Code. 15471996
Brand: Thermo Scientific Alfa Aesar H55255.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 76802-61-4
Molecular Formula C17H25F
Molecular Weight (g/mol) 248.385
MDL Number MFCD11053471
InChI Key VCIVJVKSOMDCMN-UHFFFAOYSA-N
Synonym 1-fluoro-4-4-pentylcyclohexyl benzene, trans-4'-pentylcyclohexyl-4-fluorobenzene, benzene, 1-fluoro-4-trans-4-pentylcyclohexyl, 1-fluoro-4-1s,4r-4-pentylcyclohexyl benzene, 1-fluoro-4-4-pentylcyclohexyl benzene #, benzene, 1-fluoro-4-4-pentylcyclohexyl, 1-fluoro-4-trans-4-pentylcyclohexyl benzene, trans-1-fluor-4-4-pentylcyclohexyl-benzene
PubChem CID 578937
IUPAC Name 1-fluoro-4-(4-pentylcyclohexyl)benzene
SMILES CCCCCC1CCC(CC1)C2=CC=C(C=C2)F
Specifications
Melting Point 34°C to 36°C
Quantity 5 g
Formula Weight 248.38
Percent Purity 97%
Chemical Name or Material trans-1-Fluoro-4-(4-n-pentylcyclohexyl)benzene