Learn More
trans-1,1'-(Cyclohexane-1,4-diyl)bis(N-(2-chlorobenzyl)methanamine) dihydrochloride, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 366-93-8 |
| Molecular Formula | C22H30Cl4N2 |
| MDL Number | MFCD00034973 |
| Quantity | 1 mg |
| SMILES | ClC1=CC=CC=C1CNC[C@H]2CC[C@H](CNCC3=CC=CC=C3Cl)CC2.[H]Cl.[H]Cl |
| Molecular Weight (g/mol) | 464.30 |
| Percent Purity | 98% |
| Chemical Name or Material | trans-1,1'-(Cyclohexane-1,4-diyl)bis(N-(2-chlorobenzyl)methanamine) dihydrochloride |