missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRADD Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15468-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
TRADD′s nuclear form acts as a tumor suppressor by preventing ubiquitination and degradation of isoform p19ARF/ARF of CDKN2A by TRIP12: acts by interacting with TRIP12, leading to disrupt interaction between TRIP12 and isoform p19ARF/ARF of CDKN2A. Adapter molecule for TNFRSF1A/TNFR1 that specifically associates with the cytoplasmic domain of activated TNFRSF1A/TNFR1 mediating its interaction with FADD. Overexpression of TRADD leads to two major TNF-induced responses, apoptosis and activation of NF-kappa-B.Specifications
TRADD | |
Polyclonal | |
Unconjugated | |
TRADD | |
Hs.89862, TRADD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
71609, 8717 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15628, Q3U0V2 | |
Tradd | |
TRADD Fusion Protein Ag7768 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |