Learn More
Thermo Scientific™ TracePLOT TG-BOND S GC Columns
Product Code. 10690056 Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
15 m
30 m
Diameter (Metric):
0.32 mm
0.53 mm
Unit Size:
Each
Product Code. 10690056
Brand: Thermo Scientific™ 260066060
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Choose this mid-polarity PLOT column with essentially no particle release for consistent, reproducible analysis of non-polar and polar compounds.
For consistent, reproducible analysis of nonpolar and polar compounds, Thermo Scientific™ TracePLOT TG-BOND S GC columns deliver high performance and unsurpassed stability. The columns use a midpolarity divinylbenzene 4-vinylpyridine solid phase, and the particles are incorporated into the tubing for essentially no particle release, which helps prevent damage to detection systems.
Applications:
  • Nonpolar and polar compounds
Similar To:
  • CP-PoraPLOT S
  • Supel-G45
  • Rt™-S-BOND
TRUSTED_SUSTAINABILITY
Specifications
Max. Temperature Up to 200°C
Diameter (Metric) 0.53 mm
Diameter (Metric) Inner 0.53 mm
Stationary Phase TracePLOT™ TG-BOND S
For Use With (Application) GC for Non-Polar and Mid-Polar Compounds
Film Thickness 20 μm
Length (Metric) 15 m
Polarity Mid-Polar