Learn More
Thermo Scientific™ TraceGOLD TG-35MS GC Columns
Product Code. 10279754 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
15 m
30 m
Diameter (Metric) Inner:
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.25 μm
0.5 μm
1 μm
1.5 μm
3 μm
Unit Size:
Each
Product Code. Length (Metric) Diameter (Metric) Inner Film Thickness unitSize
10279754 30 m 0.32 mm 0.5 μm Each
10505635 15 m 0.25 mm 0.25 μm Each
10534115 15 m 0.32 mm 0.25 μm Each
10016174 30 m 0.25 mm 0.25 μm Each
10167704 30 m 0.32 mm 0.25 μm Each
10733877 15 m 0.25 mm 0.5 μm Each
10639295 15 m 0.32 mm 0.5 μm Each
10545815 15 m 0.53 mm 0.5 μm Each
10371115 30 m 0.25 mm 0.5 μm Each
10781957 30 m 0.53 mm 0.5 μm Each
10621016 15 m 0.25 mm 1 μm Each
10219804 15 m 0.53 mm 1 μm Each
10629675 30 m 0.25 mm 1 μm Each
10229084 30 m 0.32 mm 1 μm Each
10713297 30 m 0.53 mm 1 μm Each
10791957 15 m 0.53 mm 1.5 μm Each
10534875 30 m 0.53 mm 1.5 μm Each
10381345 30 m 0.53 mm 3 μm Each
18 options
Product Code. 10279754
Brand: Thermo Scientific™ 260942240
Only null left
Add to basket
Add to basket
Use these columns for a higher phenyl content that provides useful elution order and retention time changes.
Thermo Scientific™ TraceGOLD TG-35MS Columns offers a higher phenyl content providing useful elution order and retention time changes.
  • Mid-polarity phase, 35% diphenyl/65% dimethyl polysiloxane
  • Equivalent to USP G42 phase
  • Similar to Rtx™-35, BD-35, HP-35, SPB-35, SPB-608
Recommended for:
  • Organochlorine pesticides and herbicides
  • Pharmaceuticals
  • PCB cogeners
  • Aroclor mixes
  • Sterols
  • Rosin acids
  • Phthalate esters
TRUSTED_SUSTAINABILITY
Specifications
Max. Temperature 300/320°C
Diameter (Metric) 0.32 mm
Diameter (Metric) Inner 0.32 mm
Product Line TraceGOLD
Stationary Phase TraceGOLD™ TG-35 MS
USP Type G42
For Use With (Application) Pharmaceuticals Analysis, GC/MS for Pharmaceuticals, GC/MS for PCB Cogeners, GC/MS for Aroclor Mixes, Sterols, Rosin Acids, and Phthalate Esters
Film Thickness 0.5 μm
Length (Metric) 30 m
Polarity Mid-Polar
Description 0.5 um Thickness; 0.32 mm ID; 30 m Length