Learn More
Thermo Scientific™ TraceGOLD TG-200MS GC Column
Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
30 m
60 m
Diameter (Metric) Inner:
0.25 mm
0.53 mm
Unit Size:
Each
Description
- Trifluoropropylmethyl polysiloxane phase resolves compounds that phenyl and cyano phases cannot
- Outstanding thermal stability and low bleed
- Suitable for use with sensitive detectors including ECD, NPD and MS
- Appropriate for use as a confirmation GC column
Specifications
Specifications
| Length (Metric) | 60 m |
| Diameter (Metric) Inner | 0.25 mm |
| USP Type | G6 |
| Stationary Phase | TraceGOLD TG-200MS |
| Film Thickness | 1 μm |