Learn More
Thermo Scientific™ TraceGOLD™ TG-1MS Guard GC Columns with SafeGuard
Product Code. 11800463 Shop All Thermo Scientific Products
Click to view available options
Diameter (Metric):
0.25 mm
0.53 mm
Particle Size:
0.25 μm
1 μm
5 μm
Unit Size:
Each
Product Code. 11800463
Brand: Thermo Scientific™ 260991425
Only null left
Add to basket
Add to basket
Extend column life with the integrated guard of the this GC column.
Extended column life with the integrated guard of the Thermo Scientific™ TraceGOLD TG-1MS GC Column with SafeGuard, which prolongs the lifetime of the analytical column.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Max. Temperature 330/350°C (Standard Columns), 430°C (Metal Columns)
Diameter (Metric) 0.25 mm
Diameter (Metric) Inner 0.25 mm
Product Line TraceGOLD
Stationary Phase TraceGOLD™ TG-1 MS
USP Type G2
For Use With (Application) GC/MS for Optimal Signal-to-Noise Ratio, Sensitivity, and MS Integrity
Particle Size 0.25 μm
Film Thickness 0.25 μm
Length (Metric) 30 m
Polarity Non-Polar