Learn More
Thermo Scientific™ TraceGOLD™ TG-1MS GC Columns
Shop All Thermo Scientific ProductsDescription
Achieve consistent results with this low-bleed, stable column. For optimal signal-to-noise ratio, sensitivity and MS integrity for a wide variety of applications, the nonpolar TraceGOLD TG-1MS GC column is an excellent choice.
Specifications
Specifications
| Max. Temperature | 330/350°C for Standard Columns; 430°C for Metal Columns |
| Diameter (Metric) | 0.2 mm |
| Diameter (Metric) Inner | 0.2 mm |
| Product Line | TraceGOLD |
| Stationary Phase | TraceGOLD™ TG-1 MS |
| USP Type | G2 |
| For Use With (Application) | GC/MS for Hydrocarbons, Solvent Impurities, PCB Congeners, and Aroclor Mixes, GC/MS for Simulated Distillation, Drugs of Abuse, Natural Gas Odorants, Essential Oils, and Pesticides |
| Film Thickness | 0.33 μm |
| Length (Metric) | 12 m |
| Polarity | Non-Polar |
| Show More |