Learn More
- Home
- Products
- Chromatography Columns and Cartridges
- GC Columns
- GC Capillary Columns
- Thermo Scientific™ TraceGOLD™ TG-1MS GC Columns
Thermo Scientific™ TraceGOLD™ TG-1MS GC Columns
Product Code. 13448008 Shop All Thermo Scientific Products| Product Code. | Length (Metric) | Diameter (Metric) Inner | Film Thickness | unitSize |
|---|---|---|---|---|
| 13448008 | 40 m | 0.15 mm | 0.15 μm | Each |
| 10436754 | 10 m | 0.1 mm | 0.1 μm | Each |
| 13468008 | 20 m | 0.15 mm | 0.15 μm | Each |
| 15870900 | 60 m | 0.32 mm | 0.5 μm | Each |
| 10744267 | 15 m | 0.25 mm | 0.25 μm | Each |
| 10096644 | 15 m | 0.32 mm | 0.25 μm | Each |
| 15820910 | 30 m | 0.53 mm | 5 μm | Each |
| 10554875 | 30 m | 0.25 mm | 0.25 μm | Each |
| 15830910 | 100 m | 0.25 mm | 0.5 μm | Each |
| 10564875 | 30 m | 0.32 mm | 0.25 μm | Each |
| 15840910 | 30 m | 0.53 mm | 3 μm | Each |
| 10006654 | 60 m | 0.25 mm | 0.25 μm | Each |
| 10429154 | 30 m | 0.25 mm | 0.1 μm | Each |
| 10127424 | 60 m | 0.32 mm | 0.25 μm | Each |
| 10137424 | 30 m | 0.25 mm | 0.5 μm | Each |
| 10426954 | 15 m | 0.32 mm | 0.5 μm | Each |
| 10197464 | 15 m | 0.53 mm | 0.5 μm | Each |
| 10270335 | 30 m | 0.25 mm | 0.5 μm | Each |
| 10280335 | 30 m | 0.32 mm | 0.5 μm | Each |
| 10544875 | 30 m | 0.53 mm | 0.5 μm | Each |
| 10006324 | 60 m | 0.25 mm | 0.5 μm | Each |
| 10016084 | 60 m | 0.32 mm | 0.5 μm | Each |
| 10046744 | 15 m | 0.25 mm | 0.5 μm | Each |
| 10628535 | 15 m | 0.32 mm | 1 μm | Each |
| 10638535 | 15 m | 0.53 mm | 1 μm | Each |
| 10407154 | 30 m | 0.25 mm | 1 μm | Each |
| 10045784 | 30 m | 0.32 mm | 1 μm | Each |
| 10055784 | 30 m | 0.53 mm | 1 μm | Each |
| 10065784 | 60 m | 0.25 mm | 1 μm | Each |
| 10477134 | 60 m | 0.32 mm | 1 μm | Each |
| 10026084 | 15 m | 0.53 mm | 1.5 μm | Each |
| 10127714 | 30 m | 0.53 mm | 1.5 μm | Each |
| 15217488 | 15 m | 0.32 mm | 3 μm | Each |
| 15227488 | 30 m | 0.32 mm | 3 μm | Each |
| 15247488 | 12 m | 0.2 mm | 0.33 μm | Each |
| 15237488 | 20 m | 0.18 mm | 0.18 μm | Each |
Description
Achieve consistent results with this low-bleed, stable column. For optimal signal-to-noise ratio, sensitivity and MS integrity for a wide variety of applications, the nonpolar TraceGOLD TG-1MS GC column is an excellent choice.
Specifications
Specifications
| Max. Temperature | 330/350°C for Standard Columns; 430°C for Metal Columns |
| Diameter (Metric) | 0.15 mm |
| Diameter (Metric) Inner | 0.15 mm |
| Product Line | TraceGOLD |
| Stationary Phase | TraceGOLD™ TG-1 MS |
| USP Type | G2 |
| For Use With (Application) | GC/MS for Hydrocarbons, Solvent Impurities, PCB Congeners, and Aroclor Mixes, GC/MS for Simulated Distillation, Drugs of Abuse, Natural Gas Odorants, Essential Oils, and Pesticides |
| Film Thickness | 0.15 μm |
| Length (Metric) | 40 m |
| Polarity | Non-Polar |
| Description | TG-1 MS 40 m x 0.15 mm x 0.15 μm |
| Show More Show Less |