Learn More
- Home
- Products
- Chromatography Columns and Cartridges
- GC Columns
- GC Capillary Columns
- Thermo Scientific™ TraceGOLD TG-1701MS GC Columns
Thermo Scientific™ TraceGOLD TG-1701MS GC Columns
Product Code. 10752727 Shop All Thermo Scientific Products| Product Code. | Length (Metric) | Diameter (Metric) Inner | Film Thickness | unitSize |
|---|---|---|---|---|
| 10752727 | 60 m | 0.32 mm | 1 μm | Each |
| 10628725 | 10 m | 0.1 mm | 0.1 μm | Each |
| 10096064 | 60 m | 0.32 mm | 1.5 μm | Each |
| 10158084 | 15 m | 0.25 mm | 0.25 μm | Each |
| 10331015 | 15 m | 0.32 mm | 0.25 μm | Each |
| 10187844 | 30 m | 0.25 mm | 0.25 μm | Each |
| 10066214 | 30 m | 0.32 mm | 0.25 μm | Each |
| 10566185 | 30 m | 0.53 mm | 0.25 μm | Each |
| 10076214 | 60 m | 0.25 mm | 0.25 μm | Each |
| 10381205 | 60 m | 0.32 mm | 0.25 μm | Each |
| 10437524 | 60 m | 0.53 mm | 0.25 μm | Each |
| 10447524 | 15 m | 0.25 mm | 0.5 μm | Each |
| 10086024 | 15 m | 0.32 mm | 0.5 μm | Each |
| 10330975 | 15 m | 0.53 mm | 0.5 μm | Each |
| 10249614 | 30 m | 0.25 mm | 0.5 μm | Each |
| 10565435 | 30 m | 0.32 mm | 0.5 μm | Each |
| 10678525 | 30 m | 0.53 mm | 0.5 μm | Each |
| 10016274 | 60 m | 0.25 mm | 0.5 μm | Each |
| 10468294 | 60 m | 0.32 mm | 0.5 μm | Each |
| 10197754 | 60 m | 0.53 mm | 0.5 μm | Each |
| 10065984 | 15 m | 0.25 mm | 1 μm | Each |
| 10108054 | 15 m | 0.32 mm | 1 μm | Each |
| 10574685 | 15 m | 0.53 mm | 1 μm | Each |
| 10732927 | 30 m | 0.25 mm | 1 μm | Each |
| 10640836 | 30 m | 0.32 mm | 1 μm | Each |
| 10649295 | 30 m | 0.53 mm | 1 μm | Each |
| 10555815 | 60 m | 0.25 mm | 1 μm | Each |
| 10406764 | 60 m | 0.53 mm | 1 μm | Each |
| 10371545 | 15 m | 0.53 mm | 1.5 μm | Each |
| 10127884 | 30 m | 0.32 mm | 1.5 μm | Each |
| 10321385 | 30 m | 0.53 mm | 1.5 μm | Each |
| 10310835 | 30 m | 0.53 mm | 3 μm | Each |
| 10610836 | 60 m | 0.53 mm | 3 μm | Each |
Description
Thermo Scientific™ TraceGOLD TG-1701MS GC Columns feature a mix of cyano and phenyl groups for increased polarity and a different elution order relative to less polar columns.
- Mid-polarity phase (14% cyanopropylphenyl, 86% dimethyl polysiloxane)
- Fully characterized for long-term reproducibility, column-to-column consistency and low bleed
- Optimal for confirmation analysis
- Equivalent to USP G46 phase
- Similar to: Rtx-1701, DB-1701, HP-1701, SPB-1701, VF-1701, CP-Sil 19 CB
Recommended for:
Alcohols
Pesticides
Oxygenates
PCB cogeners
Aroclor mixes
Specifications
Specifications
| Max. Temperature | 260/280°C |
| Diameter (Metric) | 0.32 mm |
| Diameter (Metric) Inner | 0.32 mm |
| Product Line | TraceGOLD |
| Stationary Phase | TraceGOLD™ TG-1701 MS |
| USP Type | G46 |
| For Use With (Application) | PCB Congener Analysis, GC/MS for Pesticides, GC/MS for Oxygenates, GC/MS for PCB Cogeners, GC/MS for Aroclor Mixes |
| Film Thickness | 1 μm |
| Length (Metric) | 60 m |
| Polarity | Mid-Polar |
| Description | 1 um Thickness; 0.32 mm ID; 60 m Length |
| Show More Show Less |