Learn More
Fisherbrand™ TraceableLIVE™ Subscription Coupon Codes
Shop All Fisherbrand Products
Click to view available options
Description:
1 Year Premium Subscription Coupon Code
1 Year Standard Subscription Coupon Code
2 Year Premium Subscription Coupon Code
2 Year Standard Subscription Coupon Code
Unit Size:
Each
Description
FREE - Try TraceableLIVE for free. Get Premium benefits for 30 days.
- E-mail and push alarms
- 1 user only
- 1 month rolling data storage
- Up to 3 devices can be added to the account
- Calibration due and other notifications
Standard - Equip your team to protect your sensitive items and meet most national and local regulations.
- E-mail and push alarms
- Unlimited users
- Unlimited data storage
- No limit to devices that can be added to account (each device must have subscription)
- Calibration due and other notifications
- Reports: PDF, Secured PDF, CSV and VFC
- Pre-loaded templates set up for monitoring Spex™ chemical products
Premium -Future-proof your vaccine and sample monitoring for ever-changing compliance requirements.
- E-mail and push alarms
- Unlimited users
- Unlimited data storage
- Apple Watch compatibility
- SMS text notifications included
- Voice notification escalation
- No limit to devices that can be added to account (each device must have subscription)
- Create Recurring Reports: PDF, Secured PDF, CSV and VFC
- Create your own monitoring and notification templates
- Unlimited templates
- Pre-loaded templates set up for monitoring Spex chemical products
- 21 CFR Part 11 compliance**
**Meets 21 CFR Part 11 North American Compliance and Annex 11 for Europe
Specifications
Specifications
| For Use With (Equipment) | Compatible Traceable Devices |
| Description | 2 Year Standard Subscription Coupon Code |
| Product Type | Coupon Code |
| For Use With (Application) | Cloud Monitoring |
| Includes | Card with Unique Coupon Code and QR Code |