Learn More
Thermo Scientific™ UHPLC Filters Product Code.: 11550944
Estimated Shipment: 08-10-2025
  • ONLINE EXCLUSIVE PRICE
    4060.00 NOK
    4038.34 NOK / Pack of 5
    Save 21.66 NOK
    0.53% Off
Only null left
Add to basket
Thermo Scientific™ UHPLC Filters
Product Code. p-6827001 Shop All Thermo Scientific Products
1.0 mm I.D. Replacement Filter Cartridge, 0.2 μm, Pack of 5
Click to view available options
Type:
1.0 mm I.D. Replacement Filter Cartridge, 0.2 μm
2.1 mm I.D. Replacement Filter Cartridge, 0.2 μm
UHPLC Direct Connect Filter Holder
Unit Size:
Each
Pack of 5
Product Code. 11550944
Brand: Thermo Scientific™ 22185
  • ONLINE EXCLUSIVE PRICE
    4060.00 NOK
    4038.34 NOK / Pack of 5
    Save 21.66 NOK
    0.53% Off
Only null left
Estimated Shipment: 08-10-2025
Add to basket
Only null left
Add to basket
Protects columns without compromising performance.
Sacrificing performance is not an option with Thermo Scientific™ UHPLC filters that protect columns for Hypersil GOLD™ 1.9μm and Syncronis 1.7μm columns. These UHPLC filters provide a low-volume filter cartridge design, maintain peak shape, and ensure minimal efficiency loss through dispersion.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
TRUSTED_SUSTAINABILITY
Specifications
Particle Size 0.2 μm
Type 1.0 mm I.D. Replacement Filter Cartridge, 0.2 μm
Diameter (Metric) Inner 1 mm
Product Line Thermo Scientific
Product Content Correction
Product Title
Thermo Scientific™ UHPLC Filters >