Learn More
Thermo Scientific™ TRACE™ TR-1 GC Columns Product Code.: 10496873
Estimated Shipment: 07-10-2025
  • ONLINE EXCLUSIVE PRICE
    13785.00 NOK
    13712.59 NOK / Each
    Save 72.41 NOK
    0.53% Off
Only null left
Add to basket
Thermo Scientific™ TRACE™ TR-1 GC Columns
Product Code. 10496873 Shop All Thermo Scientific Products
60 m, 0.25 mm, 1 ÎĽm, Each
Click to view available options
Length (Metric):
100 m
15 m
30 m
60 m
Diameter (Metric) Inner:
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.25 ÎĽm
0.5 ÎĽm
1 ÎĽm
1.5 ÎĽm
3 ÎĽm
5 ÎĽm
Unit Size:
Each
Product Code. 10496873
Brand: Thermo Scientific™ 260A308P
  • ONLINE EXCLUSIVE PRICE
    13785.00 NOK
    13712.59 NOK / Each
    Save 72.41 NOK
    0.53% Off
Only null left
Estimated Shipment: 07-10-2025
Add to basket
Only null left
Add to basket
Use this non-polar GC column for method development and analysis of chlorinated and nitroaromatic compounds and environmental samples.
Use the Thermo Scientific™ TRACE™ TR-1 GC column for method development and chlorinated and nitroaromatic compounds and environmental analyses. These non-polar phase, 100% dimethyl polysiloxane gas chromatography columns have a high operating temperature.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inner 0.25 mm
Product Line TRACE
Stationary Phase TR-1
Film Thickness 1 ÎĽm
Length (Metric) 60 m
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Content Correction
Product Title
Thermo Scientific™ TRACE™ TR-1 GC Columns >