Learn More
Thermo Scientific™ TRACE™ GC Columns for Pesticides
Shop All Thermo Scientific ProductsDescription
Identify organophosphate and organochlorine pesticides with high performance, sensitivity, and reproducibility using the Thermo Scientific™ TRACE™ GC Columns for Pesticides. These columns are specifically designed and tested to meet the most stringent requirements for the analysis of pesticides and herbicides.
- Low bleed decreases MS contamination
- Innovative design facilitates use at higher temperatures
- Column inertness results in minimal peak tailing and decreased breakdown of sensitive samples
- Chlorinated aromatics
- Plant sterols
- Pyrethroids
Specifications
Specifications
| Max. Temperature | 280/300°C |
| Diameter (Metric) | 0.25 mm |
| Diameter (Metric) Inner | 0.25 mm |
| Product Line | TRACE |
| Stationary Phase | Trace™ Pesticide |
| USP Type | G43 |
| For Use With (Application) | Pesticides Analysis |
| Particle Size | 0.2 μm |
| Film Thickness | 0.25 μm |
| Length (Metric) | 30 m |
| Show More |