Learn More
Invitrogen™ TRA-1-60 (Podocalyxin) Monoclonal Antibody (TRA-1-60), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
16350.00 NOK - 36070.00 NOK
Specifications
| Antigen | TRA-1-60 (Podocalyxin) |
|---|---|
| Clone | TRA-1-60 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17190682
|
Invitrogen™
14-8863-37 |
2 mg |
16350.00 NOK
2mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17100692
|
Affymetrix eBioscience
14-8863-95 |
10 mg | |||||||
Description
The TRA-1-60 antibody recognizes a protein expressed on undifferentiated human embryonic stem cells (ES), embyronal carcinoma cells (EC), and embryonic germ cells (EG). Like other stem cell specific markers, the epitope recognized by the TRA-1-60 antibody is lost upon cell differentiation. Contrary to early reports that the TRA-1-60 epitope can be destroyed by neuraminidase digestion, new strudies have shown that TRA-1-60 recognizes a neurominidase resistent antigen. The TRA-1-60 antibody is known to specifically recognize a carbohydrate epitope on a keratan sulfated glycoprotein recently identified as podocalyxin, a member of the CD34-related family of sialomucins. Podocalyxin is a transmembrane glycoprotein originally identified on epithelial glomerular cells known as podocytes, and the protein has also been implicated in the development of aggressiveness in a variety of cancers, including breast and prostate cancer.
TRA-1-60 is a cell surface antigen, expressed along with SSEA-3, SSEA-4 and TRA-1-81 in human embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). These surface markers are lost during the differentiation process. In contrast, SSEA-1 is absent in undifferentiated human stem cells but is present on the cell surface after retinoic acid mediated differentiation.Specifications
| TRA-1-60 (Podocalyxin) | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| IgM | |
| Affinity chromatography | |
| Antibody |
| TRA-1-60 | |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| TRA160; tra60 | |
| Primary | |
| 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.