Learn More
Trace Metals Standards without Mercury For Environmental Analysis, Set of 3, SPEX CertiPrep™
Product Code. 15216336 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
3 x 125 mL
Unit Size:
Pack of 3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15216336 3 x 125 mL Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15216336 Supplier SPEX Certiprep Supplier No. TMSETN
  • ONLINE EXCLUSIVE PRICE
    5545.00 NOK
    5215.99 NOK / Pack of 3
    Save 329.01 NOK
    5% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Trace Metals Ground and Wastewater Pollution Control Check Standards without Mercury for AA & ICP, 125 mL, SPEX CertiPrep™. ISO Guide 34 Certified, NIST Traceable Certified Reference Materials. Supplied with comprehensive Certificate of Analysis.
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
  • Trace Metals
  • 125 mL volume
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Compliance
GHS/OSHA Complaint
Specifications
Chemical Name or Material Trace Metals without Mercury
Name Note Set of 3 Groundwater and Wastewater Pollution Control Check Standard
Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Industry Type Wastewater Testing
Quantity 3 x 125 mL
Grade Environmental
Product intended for laboratory use only