Learn More
Invitrogen™ TR-FRET Coregulator Buffer J
Description
A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.
Specifications
Specifications
| Content And Storage | TR-FRET Coregulator Buffer J may be shipped frozen or at room temperature. It should be stored at room temperature (20-30°C). |
| Format | Liquid |
| Quantity | 25 mL |
| Shipping Condition | Approved for shipment at Room Temperature or on Wet or Dry Ice |