Learn More
TPCN1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 23758-1-AP-20UL
 View more versions of this product
Product Code. 16842435
  • 1710.33 NOK / 20µL
Description
TPCN1 may function as one of the major voltage-gated Ca2+ channel.
Specifications
Specifications
TPCN1
Polyclonal
Unconjugated
TPCN1
KIAA1169, TPC1, TPCN1, two pore segment channel 1
Rabbit
Antigen Affinity Chromatography
RUO
252972, 53373
-20°C
Liquid
Immunofluorescence, Immunocytochemistry, Western Blot
0.46 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9EQJ0, Q9ULQ1
TPCN1
TPCN1 Fusion Protein Ag20607
20 μL
Primary
Human, Mouse