Learn More
TPBG Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 29394-1-AP-150UL
 View more versions of this product
Product Code. 17571406
  • 4836.10 NOK / 150µL
Description
The TPBG antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human TPBG. This antibody recognizes human, rat, mouse antigen. The TPBG antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
TPBG
Polyclonal
Rabbit
Primary
Human, Mouse, Rat
Western Blot, Immunohistochemistry
Unconjugated
150 μL
7162