missing translation for 'onlineSavingsMsg'
Learn More

TPBG Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Product Code. 17581406 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
17581406 20 μL 20µL
17571406 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 17581406

Brand: Proteintech 293941AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The TPBG antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human TPBG. This antibody recognizes human, rat, mouse antigen. The TPBG antibody has been validated for the following applications: IHC, WB, ELISA analysis.

Specifications

Antigen TPBG
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 7162
Target Species Human, Mouse, Rat
Isotype IgG
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.