Learn More
Tocris Bioscience™ TP 472N, Tocris Bioscience™
GREENER_CHOICE
Negative control for TP 472
Brand:  Tocris Bioscience™ 5999/10
Encompass
Product Code. 15776805
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
TP 472N is a negative control for TP 472 . Inactive against other bromodomains (>20 μM against BRD9).
Active Analog also available.
Specifications
Specifications
TP 472N
10 mg
Solid
Bioactive Small Molecule
1-(8-(2-(Cyclopropylmethoxy)phenyl)pyrrolo[1,2-a]pyrimidin-6-yl)ethanone
Soluble to 100mM in DMSO and to 20mM in ethanol
Yellow
Certificates
Promotions
Promotions
Product Title
Select an issue
For Research Use Only.