Learn More
Tocris Bioscience™ TP 472N, Tocris Bioscience™
Negative control for TP 472
Brand: Tocris Bioscience™ 5999/10
Description
TP 472N is a negative control for TP 472 . Inactive against other bromodomains (>20 μM against BRD9).Active Analog also available.
Specifications
| TP 472N | |
| 10 mg | |
| Solid | |
| Bioactive Small Molecule |
| 1-(8-(2-(Cyclopropylmethoxy)phenyl)pyrrolo[1,2-a]pyrimidin-6-yl)ethanone | |
| Soluble to 100mM in DMSO and to 20mM in ethanol | |
| Yellow |
For Research Use Only.