Learn More
Tozasertib, MedChemExpress
Product Code. 16489845
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
Product Code. 16489845
Brand: Medchem Express HY10161
MedChemExpress Tozasertib (VX 680; MK-0457) is an inhibitor of Aurora A/B/C kinases with Kis of 0.6, 18, 4.6 nM, respectively.
Tozasertib (VX 680
Specifications
Chemical Name or Material Tozasertib
Physical Form Solid
CAS 639089-54-6
Color White
Quantity 250 mg
Purity Grade Notes Research
Molecular Formula C23H28N8OS
For Use With (Application) Cancer-Kinase/protease
Synonym VX 680 MK-0457
Solubility Information DMSO : ≥ 106.67 mg/mL (229.60 mM)
SMILES CC1=CC(NC2=NC(SC3=CC=C(C=C3)NC(C4CC4)=O)=NC(N5CCN(CC5)C)=C2)=NN1
Molecular Weight (g/mol) 464.59
Formula Weight 464.59
Percent Purity 98.02%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.