Learn More
Serva Electrophoresis™ Towbin Buffer, 10X
Click to view available options
Quantity:
1 L
Unit Size:
Each
Specifications
Specifications
| Chemical Name or Material | Towbin Buffer |
| Color | Colorless |
| Concentration | 10X |
| For Use With (Equipment) | For Use with Western Blotting |
| pH | 8.6 |
| Physical Form | Solution |
| Quantity | 1 L |
| Formulation | 0.25M Tris and 1.92M Glycine |
| Solubility Information | Soluble |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.