Learn More
Brady™ ToughWash™ Metal Detectable, Washdown Resistant Polyester Label Tape for M7 Printers
ToughWash™ polyester film with acrylic adhesive is a strong, print-on-demand label specially formulated for the Food and Beverage Processing industry.
2805.00 NOK - 3710.00 NOK
Specifications
| Autoclavable | Not Autoclavable |
|---|---|
| Adhesive Type | Metallic |
| Coating | None |
| Finish | Matte |
| Format | Continuous |
| Product Code | Brand | Color | Dimensions (L x W) | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Color | Dimensions (L x W) | Price | Quantity & Availability | ||||
|
17898854
|
Brady™
173458 |
White | 75 ft. x 1 in. |
2805.00 NOK
Pack of 1 |
Estimated Shipment: 04-12-2026
Log in to see stock. |
||||
|
17899224
|
Brady™
173468 |
White | 75 ft. x 2 in. |
3710.00 NOK
Pack of 1 |
In Stock
Log in to see stock. |
||||
|
17848924
|
Brady™
173459 |
Yellow | 75 ft. x 1 in. |
2805.00 NOK
Pack of 1 |
Estimated Shipment: 06-11-2026
Log in to see stock. |
||||
|
17899174
|
Brady™
173469 |
Yellow | 75 ft. x 2 in. |
3710.00 NOK
Pack of 1 |
In Stock
Log in to see stock. |
||||
Description
- Capable of being detected by commercially available metal detectors
- Label construction is tough against harsh washdown environments and withstands washdowns up to 175°F (80°C) and pressure up to 1500 PSI
- Service temperature of -40°C to +110°C (-40°F to +230°F)
- Made of B-854 polyester material
Specifications
| Not Autoclavable | |
| None | |
| Continuous | |
| Thermal Transfer | |
| Facility Identification, Food and Beverage Process Labeling, General Identification, Safety Identification | |
| No | |
| Rectangle | |
| -40°C to +110°C | |
| 0.229 mm | |
| 22.86 m |
| Metallic | |
| Matte | |
| Polyester | |
| Non-sterile | |
| M710, BMP71 | |
| 75 ft. | |
| -40°F to +230°F | |
| 0.009 in. | |
| 75 ft. |