Learn More
Heathrow Scientific™ Tough Spots™ Tube Labels, Roll
Shop All Thermo Scientific CO₂ Incubators ProductsDescription
Pre-cut peel off labels for tubes, delivered in a convenient roll format, are superior to conventional tapes and labels for use in research and clinical laboratories.
- Constructed of thin, chemically inert polyolefin with strong acrylic adhesive
- Adheres to most plastic, glass and metal mictrotubes and other items
- Labels will not peel or migrate; ideal for use in centrifuge rotors without binding or catching
- Wide assortment of colors for easy sample or researcher identification
- Round, pre-cut, peel-off labels for tube tops in convenient dispensing rolls
- 13 mm diameter spots are perfectly sized for 1.5 mL/2.0 mL tubes
- Withstands extreme environments so tubes stay clearly identified
- Heat: steam autoclaving, boiling water
- Cold: freezer temperatures to -196°C
- Organic solvents and caustic agents
- Centrifugation
Specifications
Specifications
| Adhesive Type | Acrylic |
| Color | Lavender |
| Display | Blank |
| Finish | Matte |
| Format | Roll |
| Material | Polyolefin |
| Product Type | Label |
| Sterility | Non-sterile |
| For Use With (Application) | Labeling of tubes used in freezers, boiling water, autoclave, centrifuge, ovens |
| For Use With (Equipment) | 1.5/2.0 mL Tubes |
| Show More |