Learn More
Heathrow Scientific™ Tough-Spots™ Tube Labels
Product Code. 16482015
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
16482015 White Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16482015 Supplier Heathrow Scientific™ Supplier No. HEATSPOTS44
  • ONLINE EXCLUSIVE PRICE
    460.00 NOK
    405.97 NOK / Pack of 1000
    Save 54.03 NOK
    11% Off
Estimated Shipment: 04-01-2027
Tough-Spots™ Tube Labels are easy to write PVC cryogenic labels that accept writing from most lab markers.
  • Resistant to most solvents and caustic agents
  • Adheres to most plastics, glass, and metals without cracking, peeling or degrading
  • Label sheets will not jam printers
  • Flexible polyolefin material and acrylic adhesive
  • Resistance: -196 to +150°C
  • Autoclavable
  • Disposable
Specifications
Autoclavable Autoclavable
Adhesive Type Acrylic
Color White
Finish Matte
Format Roll
Material PVC
Print Technology None
Product Type Cryogenic Label
Sterility Non-sterile
For Use With (Application) 1.5/2.0 mL Tubes
No. per Pack 1000/Pk.
Temperature Range (Metric) -196°C to +150°C
Dimensions (L x W) 1 x 1 cm
Length (English) 0.4 in.
Width (English) 0.4 in.
Width (Metric) 1 cm
Diameter (Metric) 13 mm