Learn More
DiversifiedBiotech TOUGH-SPOTS™ Roll Labels - 1.5 mL Tubes (13 mm)
Product Code. 30282284
Change view
Click to view available options
Color:
Coffee
Gold
Lavender
Maroon
Orange
Pink
Silver
Unit Size:
Pack of 1000
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30282284 Pink Pack of 1000
30282285 Coffee Pack of 1000
30282286 Gold Pack of 1000
30282273 Lavender Pack of 1000
30282347 Maroon Pack of 1000
30282298 Orange Pack of 1000
30282310 Silver Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 30282284 Supplier DiversifiedBiotech Supplier No. TSPOTS50P
  • ONLINE EXCLUSIVE PRICE
    389.00 NOK
    343.73 NOK / Pack of 1000
    Save 45.27 NOK
    11% Off
Estimated Shipment: 04-01-2027
Pre-cut peel off labels for tube tops with flexible polyolefin material
  • Circular cryogenic labels formatted for handwriting
  • Tough Spots™ on a roll are easy to write PVC cryogenic labels that accept writing from most lab markers
  • Tough Spots™ come in a variety of colors and sizes to ensure the right label for the right job
  • Adheres to most plastics (100% virgin PP tubes recommended), glass and metals without cracking, peeling or degrading
  • Flexible polyolefin material and acrylic adhesive expand and contract with temperature fluctuations
  • Label Dimensions: 0.50 in. (13 mm)
  • Roll of 1000
Specifications
Adhesive Type Peel and Stick, Acrylic
Color Pink
Finish Semi-gloss
Format Roll
Material Polyolefin
Print Technology Laser
Product Type Ultra Reflective Letter Label
Sterility Non-sterile
Length (English) 0.50 in.
Width (English) 0.50 in.
Width (Metric) 1.27 cm