Learn More
Trajan™ Autosampler Syringes for Agilent GCs
Click to view available options
Volume (Metric):
10 μL
50 μL
Unit Size:
Each
Pack of 25
Pack of 6
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Specifically designed for Agilent instruments
Specifications
Specifications
| Material | Borosilicate Glass (Barrel), PTFE (Plunger) |
| Needle Gauge | 23 and 26 G |
| Diameter (Metric) Inner Needle | 0.11 mm |
| Diameter (Metric) Outer | 6.5 mm |
| Diameter (Metric) Outer Needle | 0.63/0.47 mm |
| For Use With (Application) | Gas chromatography |
| Length (Metric) | 42 mm |
| Product Type | Gas-tight GC Autosampler Syringe |
| Model | 10F-AG-GT-0.63/0.47C |
| Quantity | 6/Pk. |
| Show More |