Learn More
WTW™ Total Nitrogen Test Kits
Product Code. 11731866
Change view
Click to view available options
Range:
0.5 to 15 mg/L
0.5 to 15 mg/L N Total
0.5 to 25 mg/L
10 to 150 mg/L
5 to 150 mg/L
Unit Size:
0.15kg
0.25kg
Each
Pack of 50
Product Code. Range unitSize
11731866 0.5 to 25 mg/L Pack of 50
11754725 10 to 150 mg/L 0.15kg
10201595 0.5 to 15 mg/L Each
11751073 0.5 to 15 mg/L N Total 0.25kg
11741866 5 to 150 mg/L Pack of 50
5 options
Product Code. 11731866
Brand: WTW™ 251995
Obtain precision results using the CombiCheck Total Nitrogen NTotal cell ready-made test kit for the determination of total nitrogen in waste water, drinking water, and surface water.
  • Variety of instrumentation for testing nitrogen in water
  • For waste water, drinking water, surface water
  • Nitrospectral Method
  • Photometric Test Methods
  • CombiCheck Total Nitrogen NTotal cell test kit
  • Safe determinations of COD and NTotal in water
TRUSTED_SUSTAINABILITY
Specifications
Analytes Chromosulfuric Acid oxidation
For Use With (Application) For waste water, drinking water, surface water
For Use With (Equipment) For Photoflex™ photometers
No. of Tests 25
Range 0.5 to 25 mg/L
Test Method Photometric
compliance-icons compliance-icons
Product Identifier
  • vario total nitrogen hydroxide LR vial
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
  • P362-Take off contaminated clothing.
  • P390-Absorb spillage to prevent material damage.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide