Learn More
Total Acid Number Standard, ASTM D664 (nominal value: 10.14 mg KOH/g), 1 x 50g, Paragon Scientific™
Product Code. 16020440
Change view
Click to view available options
Quantity:
1 x 50 g
Unit Size:
50g
Product Code. Quantity unitSize
16020440 1 x 50 g 50g
1 options
Product Code. 16020440
Brand: Paragon Scientific TAN100
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifically manufactured for the verification of analytical instruments used to determine acid number by potentiometric titration. Each standard is certified in strict accordance with international test method protocols, ASTM D664 / IP 177.
  • Acid Number (TAN) tested and certified in accordance with ASTM D664 / IP 177
  • Certified in strict accordance to ISO 17025 and ISO 17034 under our UKAS accreditation
  • Highest level of accreditation guarantee, providing the most credible certified data available worldwide
  • Fully traceable to international standards and low levels of uncertainty achieves maximum accuracy of data
  • Typically used in, but not limited to, the analysis of used oils and lubricants
  • Supplied as single volume packs or as triple packs
  • Supplied in tamper-evident glass packaging, providing assurance of sample integrity
  • Manufactured in the United Kingdom
  • 24 month shelf life
Specifications
Quantity 1 x 50 g
Nominal values quoted. Certified Values will vary from batch to batch.